Anti-UBE2J1 Mouse Monoclonal Antibody [clone: 6A12]
ABNOH00051465-M01
: Abnova
- Antibody type:Primary
- Antigen name:Ubiquitin-conjugating Enzyme E2, J1
- Antigen symbol:UBE2J1
- Clonality:Monoclonal
- Clone:6A12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:51465
- Antigen synonyms:Yeast ubiquitin-conjugating enzyme UBC6 homolog E.|UBE2J1|Ubiquitin-conjugating enzyme E2 J1|NCUBE1|NCUBE-1|UB2J1_HUMAN|HSUBC6e|Non-canonical ubiquitin-conjugating enzyme 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: UBE2J1
Clonality: Monoclonal
Clone: 6A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human