Anti-TAOK3 Mouse Monoclonal Antibody [clone: 2B8]
Catalog # ABNOH00051347-M08
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:TAO kinase 3
- Antigen symbol:TAOK3
- Clonality:Monoclonal
- Clone:2B8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:51347
- Antigen synonyms:JIK|MAP3K18|DPK
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TAOK3
Clonality: Monoclonal
Clone: 2B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human