Anti-RNF11 Mouse Polyclonal Antibody
Catalog # ABNOH00026994-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ring Finger Protein 11
- Antigen symbol:RNF11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:26994
- Antigen synonyms:Sid 1669|CGI 123,RNF11|SID1669.|RNF11_HUMAN
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RNF11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human