Order Entry
Ireland
ContactUsLinkComponent
Anti-TIMM9 Mouse Monoclonal Antibody [clone: 1D6]
Anti-TIMM9 Mouse Monoclonal Antibody [clone: 1D6]
Catalog # ABNOH00026520-M01
Supplier:  Abnova
undefined
Anti-TIMM9 Mouse Monoclonal Antibody [clone: 1D6]
Catalog # ABNOH00026520-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Translocase of Inner Mitochondrial Membrane 9
  • Antigen symbol:
    TIMM9
  • Clonality:
    Monoclonal
  • Clone:
    1D6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    26520
  • Antigen synonyms:
    TIM9A|TIM9
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: TIMM9
Clonality: Monoclonal
Clone: 1D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat