Anti-CNNM3 Mouse Polyclonal Antibody
ABNOH00026505-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Cyclin And CBS Domain divalent metal cation transport mediator 3
- Antigen symbol:CNNM3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:26505
- Antigen synonyms:ACDP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: CNNM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human