Anti-CNTNAP2 Mouse Polyclonal Antibody
Catalog # ABNOH00026047-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Contactin Associated Protein-like 2
- Antigen symbol:CNTNAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:26047
- Antigen synonyms:CASPR2|CDFE|AUTS15|PTHSL1|NRXN4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: CNTNAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human