Anti-L3MBTL1 Mouse Polyclonal Antibody
Catalog # ABNOH00026013-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:L(3)Mbt-Like 1
- Antigen symbol:L3MBTL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:26013
- Antigen synonyms:L(3)Mbt (Drosophila)-Like|H-L(3)MBT|L3MBTL|H-L(3)Mbt Protein|Lethal (3) Malignant Brain Tumor L(3)
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: L3MBTL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human