Anti-POFUT1 Rabbit Polyclonal Antibody
Catalog # ABNOH00023509-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein O-Fucosyltransferase 1
- Antigen symbol:POFUT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:23509
- Antigen synonyms:FUT12|KIAA0180|OFUCT1|DDD2|O-Fuc-T|O-FUT|GDP-fucose protein O-fucosyltransferase 1|O-FucT-1|Peptide-O-fucosyltransferase 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: POFUT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse