Order Entry
Ireland
ContactUsLinkComponent
Anti-CCT5 Mouse Monoclonal Antibody [clone: 4E5-4B1]
Anti-CCT5 Mouse Monoclonal Antibody [clone: 4E5-4B1]
  ABNOH00022948-M01
 :  Abnova
undefined
Anti-CCT5 Mouse Monoclonal Antibody [clone: 4E5-4B1]
  ABNOH00022948-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Chaperonin Containing TCP1, subunit 5 (epsilon)
  • Antigen symbol:
    CCT5
  • Clonality:
    Monoclonal
  • Clone:
    4E5-4B1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    22948
  • Antigen synonyms:
    CCTE|TCP-1-epsilon|CCT-epsilon|PNAS-102|HEL-S-69
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: CCT5
Clonality: Monoclonal
Clone: 4E5-4B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human