Anti-RAPGEF4 Mouse Monoclonal Antibody [clone: 1C11]
Catalog # ABNOH00011069-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Rap guanine nucleotide exchange factor (GEF) 4
- Antigen symbol:RAPGEF4
- Clonality:Monoclonal
- Clone:1C11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:11069
- Antigen synonyms:EPAC|CGEF2|CAMP-GEFII|Nbla00496|EPAC 2|EPAC2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: RAPGEF4
Clonality: Monoclonal
Clone: 1C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human