Anti-AHCYL1 Mouse Monoclonal Antibody [clone: 5D6]
ABNOH00010768-M05
: Abnova
- Antibody type:Primary
- Antigen name:Adenosylhomocysteinase-Like 1
- Antigen symbol:AHCYL1
- Clonality:Monoclonal
- Clone:5D6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10768
- Antigen synonyms:PRO0233|DCAL|IRBIT|PPP1R78|XPVKONA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: AHCYL1
Clonality: Monoclonal
Clone: 5D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human