Anti-SPEG Rabbit Polyclonal Antibody
ABNOH00010290-D01P
: Abnova
- Antibody type:Primary
- Antigen name:SPEG Complex locus
- Antigen symbol:SPEG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10290
- Antigen synonyms:CNM5|APEG1|APEG-1|BPEG|SPEGbeta|SPEGalpha
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SPEG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human