Anti-EFS Mouse Polyclonal Antibody
Catalog # ABNOH00010278-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:embryonal Fyn-associated substrate
- Antigen symbol:EFS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10278
- Antigen synonyms:EFS1|SIN|HEFS|CASS3|EFS2|CAS3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: EFS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human