Anti-REC8 Mouse Polyclonal Antibody
ABNOH00009985-B01P
: Abnova
- Antibody type:Primary
- Antigen name:REC8 Meiotic Recombination Protein
- Antigen symbol:REC8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9985
- Antigen synonyms:Kleisin-Alpha|Meiotic Recombination Protein REC8-Like 1|Recombination And Sister Chromatid Cohesion Protein Homolog|REC8L1|Meiotic Recombination And Sister Chromatid Cohesion Phosphoprotein Of The Rad21p Family|REC8-Like 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: REC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human