Anti-RIN1 Mouse Polyclonal Antibody
Catalog # ABNOH00009610-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ras and Rab interactor 1
- Antigen symbol:RIN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9610
- Antigen synonyms:RIN 1.|ras inhibitor|Ras and Rab interactor 1|Ras inhibitor JC99|ras inhibitor 1|ras inhibitor RIN1|RIN1_HUMAN|Ras interaction/interference protein 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human