Anti-CHST3 Mouse Monoclonal Antibody [clone: 1C4]
ABNOH00009469-M01
: Abnova
- Antibody type:Primary
- Antigen name:carbohydrate (Chondroitin 6) sulfotransferase 3
- Antigen symbol:CHST3
- Clonality:Monoclonal
- Clone:1C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9469
- Antigen synonyms:C6ST|HSD|C6ST1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CHST3
Clonality: Monoclonal
Clone: 1C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human