Anti-LY86 Mouse Polyclonal Antibody
ABNOH00009450-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Lymphocyte Antigen 86
- Antigen symbol:LY86
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9450
- Antigen synonyms:MMD-1|MD1|MD-1|dJ80N2.1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: LY86
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human