Anti-CYTH3 Mouse Monoclonal Antibody [clone: 6D3-1A9]
Catalog # ABNOH00009265-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytohesin 3
- Antigen symbol:CYTH3
- Clonality:Monoclonal
- Clone:6D3-1A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9265
- Antigen synonyms:ARNO3, GRP1, PSCD3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CYTH3
Clonality: Monoclonal
Clone: 6D3-1A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human