Anti-CTNNAL1 Mouse Monoclonal Antibody [clone: 2C11]
Catalog # ABNOH00008727-M05
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Catenin alpha-like 1
- Antigen symbol:CTNNAL1
- Clonality:Monoclonal
- Clone:2C11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8727
- Antigen synonyms:alpha-CATU|alpha-like 1|ACRP|CLLP|catenin (Cadherin-associated protein)
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CTNNAL1
Clonality: Monoclonal
Clone: 2C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human