Anti-TGM2 Mouse Monoclonal Antibody [clone: 2F4]
Catalog # ABNOH00007052-M10
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Transglutaminase 2
- Antigen symbol:TGM2
- Clonality:Monoclonal
- Clone:2F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:7052
- Antigen synonyms:transglutaminase 2|transglutaminase C|transglutaminase 2 (C polypeptide|TG2|TGase-H|C polypeptide|TGM2|TGC|protein-glutamine-gamma-glutamyltransferase|tissue transglutaminase|TGase C|protein-glutamine-gamma-glutamyltransferase)
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TGM2
Clonality: Monoclonal
Clone: 2F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human