Anti-ZEB1 Mouse Monoclonal Antibody [clone: 4C4]
Catalog # ABNOH00006935-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger E-box Binding Homeobox 1
- Antigen symbol:ZEB1
- Clonality:Monoclonal
- Clone:4C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6935
- Antigen synonyms:ZFHEP|TCF8|FECD6|PPCD3|ZFHX1A|NIL2A|TCF-8|NIL-2-A zinc finger protein|DELTAEF1|BZP|AREB6|Zinc finger E-box-binding homeobox 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ZEB1
Clonality: Monoclonal
Clone: 4C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human