Anti-SPAST Mouse Monoclonal Antibody [clone: 2F5]
ABNOH00006683-M02
: Abnova
- Antibody type:Primary
- Antigen name:Spastin
- Antigen symbol:SPAST
- Clonality:Monoclonal
- Clone:2F5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6683
- Antigen synonyms:SPG4|KIAA1083|FSP2 |OTTHUMP00000158570|ADPSP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SPAST
Clonality: Monoclonal
Clone: 2F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human