Anti-SORT1 Mouse Polyclonal Antibody
Catalog # ABNOH00006272-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Sortilin 1
- Antigen symbol:SORT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6272
- Antigen synonyms:Gp95|100 kDa NT receptor|NTR3|glycoprotein 95|sortilin|neurotensin receptor 3|LDLCQ6|NT3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SORT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human