Anti-RBBP6 Mouse Monoclonal Antibody [clone: 5A11]
Catalog # ABNOH00005930-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Retinoblastoma binding protein 6
- Antigen symbol:RBBP6
- Clonality:Monoclonal
- Clone:5A11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5930
- Antigen synonyms:PACT|SNAMA|MY038|RBQ-1|P2P-R
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: RBBP6
Clonality: Monoclonal
Clone: 5A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human