Anti-NELL1 Mouse Monoclonal Antibody [clone: 6A8]
Catalog # ABNOH00004745-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Neural EGFL like 1
- Antigen symbol:NELL1
- Clonality:Monoclonal
- Clone:6A8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4745
- Antigen synonyms:NRP1|IDH3GL
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: NELL1
Clonality: Monoclonal
Clone: 6A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human