Anti-NDUFS4 Mouse Monoclonal Antibody [clone: 3F11]
Catalog # ABNOH00004724-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:NADH Dehydrogenase (Ubiquinone) Fe-S protein 4, 18kDa (NADH-coenzyme Q reductase)
- Antigen symbol:NDUFS4
- Clonality:Monoclonal
- Clone:3F11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4724
- Antigen synonyms:CI-18|AQDQ
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: NDUFS4
Clonality: Monoclonal
Clone: 3F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human