Anti-MAGEA11 Mouse Monoclonal Antibody [clone: 3A6]
ABNOH00004110-M01A
: Abnova
- Antibody type:Primary
- Antigen name:Melanoma Antigen Family A11
- Antigen symbol:MAGEA11
- Clonality:Monoclonal
- Clone:3A6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4110
- Antigen synonyms:CT1.11|MAGE-11|MAGE11|MAGEA-11
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: MAGEA11
Clonality: Monoclonal
Clone: 3A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human