Anti-SMAD1 Mouse Monoclonal Antibody [clone: 2A1]
Catalog # ABNOH00004086-M04A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:SMAD family member 1
- Antigen symbol:SMAD1
- Clonality:Monoclonal
- Clone:2A1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4086
- Antigen synonyms:mothers against DPP homolog 1|BSP-1|Mad-related protein 1|SMAD5|mothers against decapentaplegic homolog 1|SMAD1|DWFC|MADH5|TGF-beta signaling protein 1|MAD|JV5-1|transforming growth factor-beta signaling protein 1|MAD homolog 1|hSMAD1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: SMAD1
Clonality: Monoclonal
Clone: 2A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat