Anti-HUS1 Rabbit Polyclonal Antibody
ABNOH00003364-D01
: Abnova
- Antibody type:Primary
- Antigen name:HUS1 Checkpoint Clamp Component
- Antigen symbol:HUS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3364
- Antigen synonyms:hHUS1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: HUS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human