Anti-GLA Mouse Polyclonal Antibody
ABNOH00002717-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Galactosidase Alpha
- Antigen symbol:GLA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2717
- Antigen synonyms:GLA|Alpha-galactosidase A|Alpha-D-galactoside galactohydrolase|Alpha-D-galactosidase A|GALA|Melibiase|Agalsidase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: GLA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human