Anti-DAZ1 Mouse Monoclonal Antibody [clone: 3G10]
ABNOH00001617-M01
: Abnova
- Antibody type:Primary
- Antigen name:deleted in azoospermia 1
- Antigen symbol:DAZ1
- Clonality:Monoclonal
- Clone:3G10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1617
- Antigen synonyms:SPGY|DAZ
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: DAZ1
Clonality: Monoclonal
Clone: 3G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human