Anti-DGKA Mouse Polyclonal Antibody
Catalog # ABNOH00001606-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Diacylglycerol kinase, alpha 80kDa
- Antigen symbol:DGKA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1606
- Antigen synonyms:DAGK|80 kDa diacylglycerol kinase|Diacylglycerol kinase alpha|DAG kinase alpha|DAGK1|DGK-alpha|Diglyceride kinase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: DGKA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human