Anti-CTRB1 Mouse Polyclonal Antibody
ABNOH00001504-B02P
: Abnova
- Antibody type:Primary
- Antigen name:Chymotrypsinogen B1
- Antigen symbol:CTRB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1504
- Antigen synonyms:CTRB
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: CTRB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human