Anti-CFL2 Mouse Monoclonal Antibody [clone: 6G9]
Catalog # ABNOH00001073-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cofilin 2
- Antigen symbol:CFL2
- Clonality:Monoclonal
- Clone:6G9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1073
- Antigen synonyms:NEM7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CFL2
Clonality: Monoclonal
Clone: 6G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human