Anti-CALB1 Mouse Polyclonal Antibody
ABNOH00000793-B01
: Abnova
- Antibody type:Primary
- Antigen name:Calbindin 1 28kDa
- Antigen symbol:CALB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:793
- Antigen synonyms:CALB|Calb|Calb-1|CB|Calbindin 1 28kDa|Brain-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: CALB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human