Anti-TSPO Mouse Polyclonal Antibody
Catalog # ABNOH00000706-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Translocator Protein (18kDa)
- Antigen symbol:TSPO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:706
- Antigen synonyms:peripheral-type benzodiazepine receptor|MBR|DBI|benzodiazapine receptor (peripheral)|peripheral benzodiazepine receptor|IBP|OTTHUMP00000028891|pk18 |translocator protein (18kDa) |TSPO|PTBR|BZRP|PKBS|mitochondrial benzodiazepine receptor|benzodiazepine peripheral binding site|mDRC|PBR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: TSPO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human