Anti-HBS1L Mouse Monoclonal Antibody [clone: OTI1B5]
Catalog # ORIGTA800631
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:HBS1-like (S. cerevisiae)
- Antigen symbol:HBS1L
- Clonality:Monoclonal
- Clone:OTI1B5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Form:Liquid
- Amino acid number:1 to 260
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:75.3 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions:
Type: Primary
Antigen: HBS1L (HBS1-like (S. cerevisiae))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human