Anti-CALN1 Rabbit Polyclonal Antibody
ORIGTA316588
: OriGene
- Antibody type:Primary
- Antigen name:calneuron 1
- Antigen symbol:CALN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS, 0,02% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Pk:50 µG
Recommended Dilutions: Immunochemistry: 2.5 µg/ml; Western Blot: 1-2 µg/ml
Type: Primary
Antigen: CALN1 (calneuron 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat