Order Entry
Ireland
ContactUsLinkComponent
Anti-KISS1R Rabbit Polyclonal Antibody
Anti-KISS1R Rabbit Polyclonal Antibody
  ORIGTA315927
 :  OriGene
Anti-KISS1R Rabbit Polyclonal Antibody
  ORIGTA315927
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    KISS1 receptor
  • Antigen symbol:
    KISS1R
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    internal
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Pk:
    100 µl

 

 

Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: KISS1R (KISS1 receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human