Anti-STAM Rabbit Polyclonal Antibody
Catalog # ORIGTA308964
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:SH3 domain and ITAM motif 1
- Antigen symbol:STAM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Form:Liquid
- Amino acid number:16 to 326
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7). 0,01% Thimerosal was added as a preservative.
- Molecular weight:59 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by antigen affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:500-1:3000
Type: Primary
Antigen: STAM (signal transducing adaptor molecule (SH3 domain and ITAM motif) 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human