Anti-TMEM38A Rabbit Polyclonal Antibody
Catalog # ORIGTA306888
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:transmembrane Protein 38A
- Antigen symbol:TMEM38A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS containing 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography via peptide column
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunofluorescence: 20 ?g/ml
Type: Primary
Antigen: TMEM38A (transmembrane protein 38A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat