Anti-ACE2 Rabbit Polyclonal Antibody
Catalog # ORIGTA306146
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Angiotensin I converting Enzyme 2
- Antigen symbol:ACE2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Form:Liquid
- Antigen synonyms:ACE2
- Storage buffer:PBS containing 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography via peptide column
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml
Type: Primary
Antigen: ACE2 (angiotensin I converting enzyme 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse