Anti-MFSD6 Rabbit Polyclonal Antibody
ABGEAP9932B
: Abgent
- Antibody type:Primary
- Antigen name:Major Facilitator Superfamily Domain Containing 6
- Antigen symbol:MFSD6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Gene ID:59041
- Antigen synonyms:MMR2|hMMR2
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:SQKPAEPIKRQPRSC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This Mouse Stk25 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 327-355 amino acids from the C-terminal region of mouse Stk25.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: MFSD6 (major facilitator superfamily domain containing 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human