Anti-NYX Rabbit Polyclonal Antibody
ABGEAP9483A
: Abgent
- Antibody type:Primary
- Antigen name:GRAM domain containing 3
- Antigen symbol:NYX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-Terminal
- Gene ID:65983
- Antigen synonyms:HCV NS3-transactivated protein 2|GRAM domain-containing protein 3|hepatitis C virus nonstructural protein 3 transactivating protein 2|GRAM domain-containing protein 3; GRAM3_HUMAN|Gramd3|NS3TP2.|GRAM domain containing 3|FLJ21313
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:RRKRKPAGRQQPNPC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This GRAMD3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 15-43 amino acids from the N-terminal region of human GRAMD3.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: NYX (nyctalopin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human