Anti-TPK1 Rabbit Polyclonal Antibody
Catalog # ABGEAP9142A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Thiamin Pyrophosphokinase 1
- Antigen symbol:TPK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Gene ID:56957
- Antigen synonyms:PP20|HTPK1|THMD5
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:ADSYSNGYREPPEC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This OTU7B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 757-784 amino acids from the C-terminal region of human OTU7B.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TPK1 (thiamin pyrophosphokinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse