Anti-ARHGAP26 Rabbit Polyclonal Antibody
ABGEAP8086C
: Abgent
- Antibody type:Primary
- Antigen name:Rho GTPase activating Protein 26
- Antigen symbol:ARHGAP26
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Center (200–300 aa)
- Cross adsorption:No
- Gene ID:338674
- Antigen synonyms:OR11-10
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:KKALANVISRKRTC
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Immunogen:This OR5F1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 283-310 amino acids from the C-terminal region of human OR5F1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: ARHGAP26 (Rho GTPase activating protein 26)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse