Anti-CAMKK1 Rabbit Polyclonal Antibody
ABGEAP7768A
: Abgent
- Antibody type:Primary
- Antigen name:Calcium/Calmodulin-Dependent Protein Kinase Kinase 1, alpha
- Antigen symbol:CAMKK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:MGC34095|Ca2+/calmodulin-dependent protein kinase kinase alpha|CaM-kinase kinase 1|CaMKK 1|Calcium/calmodulin-dependent protein kinase kinase alpha|Calcium/calmodulin dependent protein kinase kinase alpha|CaMKK|CaMKK alpha|Calcium/calmodulin dependent protein kinase kinase 1 alpha|DKFZp761M0423|CaM-kinase IV kinase|Camkk1|Calcium/calmodulin dependent protein kinase kinase|CAMKK alpha protein|KKCC1_HUMAN|CaM-KK alpha|CAMKKA|Calcium/calmodulin-dependent protein kinase kinase 1|CaM-kinase kinase alpha|CaM-KK 1|CaM kinase kinase alpha|OTTMUSP00000006419.|Calcium/calmodulin dependent protein kinase kinase 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: CAMKK1 (calcium/calmodulin-dependent protein kinase kinase 1, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human