Anti-PBP (PPBP) Rabbit Polyclonal Antibody
Catalog # ABGEAP7556B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Pro-Platelet Basic Protein
- Antigen symbol:PPBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Antigen synonyms:Beta-TG|CTAPIII|SCYB7|CTAP3|TC2|TGB|CTAP-III|MDGF|LDGF|PBP|THBGB1|LA-PF4|THBGB|TC1|TGB1|NAP-2|CXCL7|B-TG1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: PBP (PPBP)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human