Anti-MAPK7 Rabbit Polyclonal Antibody
Catalog # ABGEAP7504A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 7
- Antigen symbol:MAPK7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:ERK4|ERK5|PRKM7|BMK1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MAPK7 (mitogen-activated protein kinase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human