Anti-DCXR Rabbit Polyclonal Antibody
Catalog # ABGEAP7443C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Dicarbonyl/L-xylulose reductase
- Antigen symbol:DCXR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Antigen synonyms:PNTSU|Kidney dicarbonyl reductase|Carbonyl reductase II|kiDCR|HCR2|SDR20C1|KIDCR|Sperm surface protein P34H|XR|L-xylulose reductase|HCRII|P34H|Dicarbonyl/L-xylulose reductase|DCR
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: DCXR (dicarbonyl/L-xylulose reductase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse